missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Glypican 5 Monoclonal Antibody (K70048_12B8), Invitrogen™
Description
Sequence of this protein is as follows: EDIVKSYTQR VVGNGIKAQS GNPEVKVKGI DPVINQIIDK LKHVVQLLQG RSPKPDKWEL LQLGSGGGMV EQVSGDCDDE DGC
Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation. [provided by RefSeq, Jul 2008].
Specifications
Specifications
| Antigen | Glypican 5 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | K70048_12B8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide |
| Gene | GPC5 |
| Gene Accession No. | P78333 |
| Gene Alias | A230034F01Rik; bA93M14.1; glypican 5; glypican proteoglycan 5; glypican-5; Gpc5; Secreted glypican-5 |
| Show More |