missing translation for 'onlineSavingsMsg'
Learn More

Glyphosate Standard, 1000μg/mL in distilled water, Restek

Catalog No. 06610556
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06610556 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06610556 Supplier Chromatographic Supplier No. RK 32426

Chemical Identifiers

CAS 1071-83-6
Molecular Formula C3H8NO5P
Molecular Weight (g/mol) 169.07
MDL Number MFCD00055350
InChI Key XDDAORKBJWWYJS-UHFFFAOYSA-N
PubChem CID 3496
ChEBI CHEBI:27744
IUPAC Name 2-[(phosphonomethyl)amino]acetic acid
SMILES OC(=O)CNCP(O)(O)=O

Specifications

Solvent or Matrix Water
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL in distilled water
Chemical Name or Material Glyphosate