Learn More
Glyoxime, 98+%, moistened with ca 20% water
CAS: 557-30-2 | C2H4N2O2 | 88.066 g/mol
Supplier: Thermo Scientific Chemicals L0513018
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Glyoxime | |
| ∽169°C (decomposition) | |
| C2H4N2O2 | |
| 50 g | |
| 1699539 | |
| RUFIRPVAEJIIIS-OWOJBTEDSA-N | |
| N-[(E)-2-nitrosoethenyl]hydroxylamine | |
| 5354753 | |
| ≥98% |
| 557-30-2 | |
| moistened with ≈20% water | |
| MFCD00013942 | |
| UN2926 | |
| glyoxime, ethanedial, dioxime, ethanedione dioxime, pik-off, glyoxaldioxime, glyoxal dioxime, ethanedial dioxime, glyoxal-dioxime, dioxime ethanedial, glyoxal, dioxime | |
| C(=CN=O)NO | |
| 88.066 | |
| 88.07 |
Chemical Identifiers
| 557-30-2 | |
| 88.066 | |
| RUFIRPVAEJIIIS-OWOJBTEDSA-N | |
| 5354753 | |
| C(=CN=O)NO |
| C2H4N2O2 | |
| MFCD00013942 | |
| glyoxime, ethanedial, dioxime, ethanedione dioxime, pik-off, glyoxaldioxime, glyoxal dioxime, ethanedial dioxime, glyoxal-dioxime, dioxime ethanedial, glyoxal, dioxime | |
| N-[(E)-2-nitrosoethenyl]hydroxylamine |
Safety and Handling
GHS H Statement
H228-H301-H312-H315-H319-H335
Flammable solid.
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H228-H301-H312-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, TOXIC, ORGANIC, N.O.S.
EINECSNumber : 209-168-1
RTECSNumber : MD2850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only