accessibility menu, dialog, popup

UserName

CAS RN 557-30-2

IUPAC Name: N-[(1E)-2-nitrosoethenyl]hydroxylamine
Synonyms: N-[(1E)-2-nitrosoethenyl]hydroxylamine
Molecular Weight (g/mol): 88.07
Molecular Formula: C2H4N2O2
InChi Key: RUFIRPVAEJIIIS-OWOJBTEDSA-N
SMILES: ON\C=C\N=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results