missing translation for 'onlineSavingsMsg'
Learn More

Glyoxime, 98+%, moistened with ca 20% water

Catalog No. AAL0513009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0513009 10 g
AAL0513018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0513009 Supplier Thermo Scientific Chemicals Supplier No. L0513009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 557-30-2 | C2H4N2O2 | 88.066 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 557-30-2
Molecular Formula C2H4N2O2
Molecular Weight (g/mol) 88.066
MDL Number MFCD00013942
InChI Key RUFIRPVAEJIIIS-OWOJBTEDSA-N
Synonym glyoxime, ethanedial, dioxime, ethanedione dioxime, pik-off, glyoxaldioxime, glyoxal dioxime, ethanedial dioxime, glyoxal-dioxime, dioxime ethanedial, glyoxal, dioxime
PubChem CID 5354753
IUPAC Name N-[(E)-2-nitrosoethenyl]hydroxylamine
SMILES C(=CN=O)NO

Specifications

Melting Point ∽169°C (decomposition)
Quantity 10 g
UN Number UN2926
Beilstein 1699539
Formula Weight 88.07
Percent Purity ≥98%
Assay moistened with ≈20% water
Chemical Name or Material Glyoxime
Hazard Category H228-H301-H312-H315-H319-H335
Hazard Statement GHS H Statement
H228-H301-H312-H315-H319-H335
Flammable solid.
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, TOXIC, ORGANIC, N.O.S.
EINECSNumber 209-168-1
RTECSNumber MD2850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only