missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Glyoxal, pure, 40 wt.% solution in water

Supplier:  Thermo Scientific™ 156220250

Catalog No. AC156220250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Glyoxal
107-22-2 , 7732-18-5
104.0°C
Metal Drum
HCOCHO
25L
01,413
15, 4544
Solubility in water: miscible.
C(=O)C=O
58.04
8 mPa.s (20°C)
58.04
Pure
40 wt.% Solution in Water
1.2650g/mL
>104°C
C2H2O2
MFCD00006957
01, 759
1.265
glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40
LEQAOMBKQFMDFZ-UHFFFAOYSA-N
oxaldehyde
7860
CHEBI:34779
39 to 41% (Titrimetry other)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only