missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyoxal (39% in Water, ca. 8.8mol/L), TCI America™
$34.18 - $55.07
Chemical Identifiers
| CAS | 107-22-2 |
|---|---|
| Molecular Formula | C2H2O2 |
| Molecular Weight (g/mol) | 58.036 |
| MDL Number | MFCD00006957 |
| InChI Key | LEQAOMBKQFMDFZ-UHFFFAOYSA-N |
| Synonym | glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40 |
| PubChem CID | 7860 |
| ChEBI | CHEBI:34779 |
| IUPAC Name | oxaldehyde |
| SMILES | C(=O)C=O |
Chemical Identifiers
| 107-22-2 | |
| 58.036 | |
| LEQAOMBKQFMDFZ-UHFFFAOYSA-N | |
| 7860 | |
| oxaldehyde |
| C2H2O2 | |
| MFCD00006957 | |
| glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40 | |
| CHEBI:34779 | |
| C(=O)C=O |
Specifications
| Clear Liquid at 20°C | |
| Yellow | |
| 25 g | |
| MFCD00006957 | |
| LEQAOMBKQFMDFZ-UHFFFAOYSA-N | |
| oxaldehyde | |
| 7860 | |
| 58.04 |
| 107-22-2 | |
| 2.1 to 2.7 | |
| C2H2O2 | |
| glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40 | |
| C(=O)C=O | |
| 58.036 | |
| CHEBI:34779 | |
| Glyoxal (39% in Water, ca. 8.8mol/L) |
Safety and Handling
EINECSNumber : (2)-0510
RTECSNumber : MD2700000
TSCA : Yes