missing translation for 'onlineSavingsMsg'
Learn More

Glyoxal (39% in Water, ca. 8.8mol/L), TCI America™

Supplier:  TCI America G0152500G

 View more versions of this product

Catalog No. G0152500G


Only null left

Description

Description

Specifications

Specifications

Clear Liquid at 20°C
107-22-2
2.1 to 2.7
C2H2O2
glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40
C(=O)C=O
58.036
CHEBI:34779
Glyoxal (39% in Water, ca. 8.8mol/L)
Yellow
500 g
MFCD00006957
LEQAOMBKQFMDFZ-UHFFFAOYSA-N
oxaldehyde
7860
58.04

Chemical Identifiers

107-22-2
58.036
LEQAOMBKQFMDFZ-UHFFFAOYSA-N
7860
oxaldehyde
C2H2O2
MFCD00006957
glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40
CHEBI:34779
C(=O)C=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0510

RTECSNumber : MD2700000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions