missing translation for 'onlineSavingsMsg'
Learn More

Glycyl-L-tryptophan Hydrate 98.0+%, TCI America™

Catalog No. G0144100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0144100MG 100 mg
G01441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0144100MG Supplier TCI America Supplier No. G0144100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2390-74-1
Molecular Formula C13H15N3O3
Molecular Weight (g/mol) 261.28
MDL Number MFCD00180733
InChI Key AJHCSUXXECOXOY-UHFFFAOYNA-N
Synonym H-Gly-Trp-OH
PubChem CID 92181
ChEBI CHEBI:73921
IUPAC Name 2-(2-aminoacetamido)-3-(1H-indol-3-yl)propanoic acid
SMILES NCC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Quantity 100 mg
Formula Weight 261.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Glycyl-L-tryptophan Hydrate