missing translation for 'onlineSavingsMsg'
Learn More

Glycyl-L-threonine Dihydrate, TCI America™

Catalog No. p-7130157
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
G01431G 1 g
1 options

Catalog No. G01431G

Supplier: TCI America G01431G

Only null left

Chemical Identifiers

CAS 7093-70-1
Molecular Formula C6H12N2O4
Molecular Weight (g/mol) 176.17
MDL Number MFCD00038176
InChI Key OLIFSFOFKGKIRH-WUJLRWPWSA-N
Synonym H-Gly-Thr-OH
PubChem CID 111257
ChEBI CHEBI:73920
IUPAC Name (2S,3R)-2-(2-aminoacetamido)-3-hydroxybutanoic acid
SMILES C[C@@H](O)[C@H](NC(=O)CN)C(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 176.17
Physical Form Crystalline Powder
Chemical Name or Material Glycyl-L-threonine Dihydrate