missing translation for 'onlineSavingsMsg'
Learn More

Glycyl-L-phenylalanine 98.0+%, TCI America™

Catalog No. p-7130155
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0136100MG 100 mg
G01361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0136100MG Supplier TCI America Supplier No. G0136100MG

Chemical Identifiers

CAS 3321-03-7
Molecular Formula C11H14N2O3
Molecular Weight (g/mol) 222.24
MDL Number MFCD00065110
InChI Key JBCLFWXMTIKCCB-UHFFFAOYNA-N
Synonym H-Gly-Phe-OH
PubChem CID 92953
ChEBI CHEBI:73912
IUPAC Name (2S)-2-[(2-aminoacetyl)amino]-3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)CC(C(=O)O)NC(=O)CN

Specifications

Color White
Quantity 100 mg
Formula Weight 222.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Glycyl-L-phenylalanine
TSCA No
Recommended Storage Refrigerator