missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycyl-L-glutamic Acid 98.0+%, TCI America™
$57.75 - $257.97
Chemical Identifiers
| CAS | 7412-78-4 |
|---|---|
| Molecular Formula | C7H12N2O5 |
| Molecular Weight (g/mol) | 204.182 |
| MDL Number | MFCD00008128 |
| InChI Key | IEFJWDNGDZAYNZ-BYPYZUCNSA-N |
| Synonym | glycyl-l-glutamic acid, n-glycylglutamic acid, gly-glu, n-glycyl-l-glutamic acid, s-2-2-aminoacetamido pentanedioic acid, 2s-2-2-aminoacetamido pentanedioic acid, l-glutamic acid, n-glycyl, glycyl-l-glutamicacid, gly-l-glu, gly-l-glu-oh |
| PubChem CID | 99278 |
| ChEBI | CHEBI:73801 |
| IUPAC Name | (2S)-2-[(2-aminoacetyl)amino]pentanedioic acid |
| SMILES | C(CC(=O)O)C(C(=O)O)NC(=O)CN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
G0123100MG
|
TCI America
G0123100MG |
100 mg |
Each for $57.75
|
|
|||||
|
G01231G
|
TCI America
G01231G |
1 g |
Each for $257.97
|
|
|||||
Chemical Identifiers
| 7412-78-4 | |
| 204.182 | |
| IEFJWDNGDZAYNZ-BYPYZUCNSA-N | |
| 99278 | |
| (2S)-2-[(2-aminoacetyl)amino]pentanedioic acid |
| C7H12N2O5 | |
| MFCD00008128 | |
| glycyl-l-glutamic acid, n-glycylglutamic acid, gly-glu, n-glycyl-l-glutamic acid, s-2-2-aminoacetamido pentanedioic acid, 2s-2-2-aminoacetamido pentanedioic acid, l-glutamic acid, n-glycyl, glycyl-l-glutamicacid, gly-l-glu, gly-l-glu-oh | |
| CHEBI:73801 | |
| C(CC(=O)O)C(C(=O)O)NC(=O)CN |
Specifications
| 7412-78-4 | |
| White | |
| MFCD00008128 | |
| glycyl-l-glutamic acid, n-glycylglutamic acid, gly-glu, n-glycyl-l-glutamic acid, s-2-2-aminoacetamido pentanedioic acid, 2s-2-2-aminoacetamido pentanedioic acid, l-glutamic acid, n-glycyl, glycyl-l-glutamicacid, gly-l-glu, gly-l-glu-oh | |
| C(CC(=O)O)C(C(=O)O)NC(=O)CN | |
| 204.182 | |
| CHEBI:73801 | |
| ≥98.0% (T) | |
| Glycyl-L-glutamic Acid |
| 158°C | |
| C7H12N2O5 | |
| 100 mg | |
| IEFJWDNGDZAYNZ-BYPYZUCNSA-N | |
| (2S)-2-[(2-aminoacetyl)amino]pentanedioic acid | |
| 99278 | |
| 204.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer