missing translation for 'onlineSavingsMsg'
Learn More

Glycyl-L-alanine 98.0+%, TCI America™

Supplier:  TCI America G01131G

 View more versions of this product

Catalog No. G01131G


Only null left

Description

Description

Specifications

Specifications

Glycyl-L-alanine
White
MFCD00065112
VPZXBVLAVMBEQI-VKHMYHEASA-N
(2S)-2-[(2-aminoacetyl)amino]propanoic acid
146.146
CHEBI:73855
≥98.0% (T)
3695-73-6
C5H10N2O3
H-Gly-Ala-OH
CC(C(=O)O)NC(=O)CN
1 g
1551643
146.15
Crystalline Powder

Chemical Identifiers

3695-73-6
146.146
VPZXBVLAVMBEQI-VKHMYHEASA-N
1551643
(2S)-2-[(2-aminoacetyl)amino]propanoic acid
C5H10N2O3
MFCD00065112
H-Gly-Ala-OH
CHEBI:73855
CC(C(=O)O)NC(=O)CN
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions