missing translation for 'onlineSavingsMsg'
Learn More

Glycyl-L-alanine 98.0+%, TCI America™

Catalog No. p-7130148
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01131G 1 g
G01135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G01131G Supplier TCI America Supplier No. G01131G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3695-73-6 | C5H10N2O3 | 146.146 g/mol

Chemical Identifiers

CAS 3695-73-6
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.146
MDL Number MFCD00065112
InChI Key VPZXBVLAVMBEQI-VKHMYHEASA-N
Synonym H-Gly-Ala-OH
PubChem CID 1551643
ChEBI CHEBI:73855
IUPAC Name (2S)-2-[(2-aminoacetyl)amino]propanoic acid
SMILES CC(C(=O)O)NC(=O)CN

Specifications

Color White
Quantity 1 g
Formula Weight 146.15
Percent Purity ≥98.0% (T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material Glycyl-L-alanine