missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycyl-D-leucine 99.0+%, TCI America™
$29.83 - $139.81
Chemical Identifiers
| CAS | 688-13-1 |
|---|---|
| Molecular Formula | C8H16N2O3 |
| Molecular Weight (g/mol) | 188.23 |
| MDL Number | MFCD00066050 |
| InChI Key | DKEXFJVMVGETOO-UHFFFAOYNA-N |
| Synonym | H-Gly-D-Leu-OH |
| PubChem CID | 1715077 |
| IUPAC Name | 2-(2-aminoacetamido)-4-methylpentanoic acid |
| SMILES | CC(C)CC(NC(=O)CN)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
G0130100MG
|
TCI America
G0130100MG |
100 mg |
Each for $29.83
|
|
|||||
|
G01301G
|
TCI America
G01301G |
1 g |
Each for $139.81
|
|
|||||
Chemical Identifiers
| 688-13-1 | |
| 188.23 | |
| DKEXFJVMVGETOO-UHFFFAOYNA-N | |
| 1715077 | |
| CC(C)CC(NC(=O)CN)C(O)=O |
| C8H16N2O3 | |
| MFCD00066050 | |
| H-Gly-D-Leu-OH | |
| 2-(2-aminoacetamido)-4-methylpentanoic acid |
Specifications
| 234°C | |
| White | |
| MFCD00066050 | |
| DKEXFJVMVGETOO-UHFFFAOYNA-N | |
| 2-(2-aminoacetamido)-4-methylpentanoic acid | |
| 188.23 | |
| 188.23 | |
| Crystalline Powder |
| 688-13-1 | |
| C8H16N2O3 | |
| H-Gly-D-Leu-OH | |
| CC(C)CC(NC(=O)CN)C(O)=O | |
| 100 mg | |
| 1715077 | |
| ≥99.0% (T) | |
| Glycyl-D-leucine |
Safety and Handling
TSCA : No