accessibility menu, dialog, popup

UserName

CAS RN 688-13-1

H3CCH3NHONH2OHO

CAS RN 688-13-1

IUPAC Name: 2-(2-aminoacetamido)-4-methylpentanoic acid
Synonyms: 2-(2-aminoacetamido)-4-methylpentanoic acid
Molecular Weight (g/mol): 188.23
Molecular Formula: C8H16N2O3
InChi Key: DKEXFJVMVGETOO-UHFFFAOYNA-N
SMILES: CC(C)CC(NC(=O)CN)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)