missing translation for 'onlineSavingsMsg'
Learn More

Glycyl-4-aminobutyric Acid 95.0+%, TCI America™

Catalog No. G01151G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G01151G Supplier TCI America Supplier No. G01151G
Only null left

Chemical Identifiers

CAS 32595-49-6
Molecular Formula C6H12N2O3
Molecular Weight (g/mol) 160.173
MDL Number MFCD00056123
InChI Key FNSFAHRUVVKKMM-UHFFFAOYSA-N
PubChem CID 99281
IUPAC Name 4-[(2-aminoacetyl)amino]butanoic acid
SMILES C(CC(=O)O)CNC(=O)CN

Specifications

Melting Point 195°C
Color White-Yellow
Quantity 1 g
Formula Weight 160.17
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Glycyl-4-aminobutyric Acid