missing translation for 'onlineSavingsMsg'
Learn More

Glycoluril 98.0+%, TCI America™

Catalog No. G024325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G024325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G024325G Supplier TCI America Supplier No. G024325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 496-46-8
Molecular Formula C4H6N4O2
Molecular Weight (g/mol) 142.118
MDL Number MFCD00005539
InChI Key VPVSTMAPERLKKM-UHFFFAOYSA-N
Synonym glycoluril, acetyleneurea, acetylenediureine, acetylenediurea, glyoxalbiuret, glyoxaldiureine, acetylene carbamide, diurea glyoxalate, tetrahydroimidazo 4,5-d imidazole-2,5 1h,3h-dione, glyoxaldiurene
PubChem CID 62347
ChEBI CHEBI:42946
IUPAC Name 1,3,3a,4,6,6a-hexahydroimidazo[4,5-d]imidazole-2,5-dione
SMILES C12C(NC(=O)N1)NC(=O)N2

Specifications

Color White
Quantity 25 g
Formula Weight 142.12
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Glycoluril