accessibility menu, dialog, popup

UserName

CAS RN 496-46-8

IUPAC Name: octahydro-[1,3]diazolo[4,5-d]imidazole-2,5-dione
Synonyms: glycoluril glyoxaldiureine diurea glyoxalate glyoxalbiuret glycoluril (8CI) acetylenediureine glyoxaldiurene
Molecular Weight (g/mol): 142.12
Molecular Formula: C4H6N4O2
InChi Key: VPVSTMAPERLKKM-UHFFFAOYSA-N
SMILES: O=C1NC2NC(=O)NC2N1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)