missing translation for 'onlineSavingsMsg'
Learn More

Glycoluril, 97%

Catalog No. AC120130250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC120130250 25 g
AC120131000 100 g
AC120135000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC120130250 Supplier Thermo Scientific Chemicals Supplier No. 120130250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 496-46-8 | C4H6N4O2 | 142.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 496-46-8
Molecular Formula C4H6N4O2
Molecular Weight (g/mol) 142.12
InChI Key VPVSTMAPERLKKM-UHFFFAOYSA-N
Synonym glycoluril, acetyleneurea, acetylenediureine, acetylenediurea, glyoxalbiuret, glyoxaldiureine, acetylene carbamide, diurea glyoxalate, tetrahydroimidazo 4,5-d imidazole-2,5 1h,3h-dione, glyoxaldiurene
PubChem CID 62347
ChEBI CHEBI:42946
IUPAC Name 1,3,3a,4,6,6a-hexahydroimidazo[4,5-d]imidazole-2,5-dione
SMILES C12C(NC(=O)N1)NC(=O)N2

Specifications

Melting Point >300.0°C
Boiling Point 862.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (N)
Quantity 25 g
Beilstein 26,441
Merck Index 15,88
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in warm mineral acids and in warm ammonia
Formula Weight 142.12
Percent Purity 97%
Chemical Name or Material Glycoluril
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 207-821-5

RUO – Research Use Only