missing translation for 'onlineSavingsMsg'
Learn More

Glycolide 98.0+%, TCI America™

Catalog No. G04675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G04675G 5 g
G046725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G04675G Supplier TCI America Supplier No. G04675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 502-97-6
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00081108
InChI Key RKDVKSZUMVYZHH-UHFFFAOYSA-N
Synonym glycolide, glycollide, unii-yrz676pgu6, p-dioxane-2,5-dione, yrz676pgu6, acmc-1aybf, 1,4-dioxan-2,5-dione, 2,5-dioxo-1,4-dioxane, p-dioxane-2,5-dione 8ci
PubChem CID 65432
IUPAC Name 1,4-dioxane-2,5-dione
SMILES O=C1COC(=O)CO1

Specifications

Melting Point 84°C
Color White
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Boiling Point 130°C
Formula Weight 116.07
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material Glycolide
EINECSNumber (5)-6815&(5)-6865
TSCA Yes
Recommended Storage Freezer (-20 °C)