accessibility menu, dialog, popup

UserName

CAS RN 502-97-6

IUPAC Name: 1,4-dioxane-2,5-dione
Synonyms: 1,4-dioxane-2,5-dione
Molecular Weight (g/mol): 116.07
Molecular Formula: C4H4O4
InChi Key: RKDVKSZUMVYZHH-UHFFFAOYSA-N
SMILES: O=C1COC(=O)CO1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)