missing translation for 'onlineSavingsMsg'
Learn More

Glycolic Acid, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019609
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019609 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019609 Supplier Merck Emd Millipore Supplier No. PHR14271G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Glycolic acid is an α-hydroxy acid with exfoliating and anti-ageing properties. It is useful in dermatology, medical, cosmetics, and pharmaceutical applications.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 79-14-1
Linear Formula HOCH2COOH
MDL Number MFCD00004312
Melting Point 75°C to 80°C
Synonym Glycolic acid; Hydroxyacetic acid
Quantity 1 g
Formula Weight 76.05
Grade Certified Reference Material