Learn More
Glycolic Acid, Certified Reference Material, MilliporeSigma™ Supelco™
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Supplier:  Merck Emd Millipore 9481510G
 View more versions of this product
Catalog No. 111017918
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Specifications
Specifications
Bottomless Glass Bottle with Fused Cone
TraceCERT
HOCH2COOH
75°C to 80°C
10 g
TraceCERT™ Analytical Standard
79-14-1
Neat
MFCD00004312
Hydroxyacetic acid
76.05
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC