Learn More
Glycolic acid, 99%
CAS: 79-14-1 | C2H4O3 | 76.051 g/mol
Supplier: Thermo Scientific Chemicals 154511000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Glycolic acid | |
| 79-14-1 | |
| White | |
| C2H4O3 | |
| 03, 228 | |
| glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech | |
| AEMRFAOFKBGASW-UHFFFAOYSA-N | |
| 2-hydroxyacetic acid | |
| 757 | |
| 76.04 | |
| 100 g | |
| Conforms to Ref | |
| Adhering Crystals |
| 72.0°C to 80.0°C | |
| 100.0°C | |
| 99% | |
| HOCH2CO2H | |
| 15, 4534 | |
| Solubility in water: soluble in water. Other solubilities: soluble in alcohol, ether, acetic acid and acetone | |
| C(C(=O)O)O | |
| 76.051 | |
| CHEBI:17497 | |
| 99% | |
| >300°C | |
| Glass bottle | |
| 8.1mmHg at 80°C |
Chemical Identifiers
| 79-14-1 | |
| 76.051 | |
| glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech | |
| CHEBI:17497 | |
| C(C(=O)O)O |
| C2H4O3 | |
| AEMRFAOFKBGASW-UHFFFAOYSA-N | |
| 757 | |
| 2-hydroxyacetic acid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage. Harmful if inhaled.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Do not breathe dust/fume/gas/mist/vapors/spray.
Danger
RUO – Research Use Only