missing translation for 'onlineSavingsMsg'
Learn More

Glycolic acid, 99%

Catalog No. p-4510276
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Packaging:
Glass Bottle
Catalog No. Quantity Packaging
AC154510250 25 g Glass Bottle
AC154511000 100 g Glass Bottle
AC154515000 500 g Glass Bottle
3 options

Catalog No. AC154510250

Supplier: Thermo Scientific Chemicals 154510250

Only null left

CAS: 79-14-1 | C2H4O3 | 76.051 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79-14-1
Molecular Formula C2H4O3
Molecular Weight (g/mol) 76.051
InChI Key AEMRFAOFKBGASW-UHFFFAOYSA-N
Synonym glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech
PubChem CID 757
ChEBI CHEBI:17497
IUPAC Name 2-hydroxyacetic acid
SMILES C(C(=O)O)O

Specifications

Melting Point 72.0°C to 80.0°C
Boiling Point 100.0°C
Color White
Assay Percent Range 99%
Linear Formula HOCH2CO2H
Beilstein 03, 228
Merck Index 15, 4534
Formula Weight 76.04
Percent Purity 99%
Quantity 25 g
Flash Point >300°C
Infrared Spectrum Conforms to Ref
Packaging Glass Bottle
Physical Form Adhering Crystals
Vapor Pressure 8.1mmHg at 80°C
Chemical Name or Material Glycolic acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement Causes severe skin burns and eye damage. Harmful if inhaled.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

RUO – Research Use Only