missing translation for 'onlineSavingsMsg'
Learn More

Glycolic Acid, 70%, High Purity, Spectrum™ Chemical

Catalog No. 18603024
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603024 100 mL Amber Glass Bottle
18603026 500 mL Amber Glass Bottle
18603027 6 x 500 mL Amber Glass Bottle
18603028 4 L Amber Glass Bottle
18603029 4 x 4 L Amber Glass Bottle
18603025 20 L Poly Drum
18603030 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18603024 Supplier Spectrum Chemical Mfg Cor Supplier No. G1059100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

G1059, 79-14-1, C2H4O3

Spectrum™ Chemical Glycolic Acid, 70 Percent, High Purity , is slightly stronger than acetic acid. It is often included into emulsion polymers, solvents and additives for ink and paint in order to improve flow properties and impart gloss. It is used in surface treatment products that increase the coefficient of friction on tile flooring. Glycolic acid is a useful synthetic organic intermediate for oxidation-reduction, esterification and long chain polymerizations.

Chemical Identifiers

CAS 79-14-1 , 7732-18-5
Molecular Formula C2H4O3
Molecular Weight (g/mol) 76.05
InChI Key AEMRFAOFKBGASW-UHFFFAOYSA-N
IUPAC Name 2-hydroxyacetic acid
SMILES OCC(O)=O

Specifications

CAS Max % 70%, 30%
Quantity 100 mL
Packaging Amber Glass Bottle
UN Number UN3265
Grade High Purity