missing translation for 'onlineSavingsMsg'
Learn More

Glycocyamine 97.0+%, TCI America™

Catalog No. G016725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G016725G 25 g
G0167500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G016725G Supplier TCI America Supplier No. G016725G

Chemical Identifiers

CAS 352-97-6
Molecular Formula C3H7N3O2
Molecular Weight (g/mol) 117.108
MDL Number MFCD00004278
InChI Key BPMFZUMJYQTVII-UHFFFAOYSA-N
Synonym glycocyamine, guanidinoacetic acid, guanidineacetic acid, guanidoacetic acid, n-amidinoglycine, 2-guanidinoacetic acid, betacyamine, guanyl glycine, betasyamine, glykocyamin
PubChem CID 763
ChEBI CHEBI:16344
IUPAC Name 2-(diaminomethylideneamino)acetic acid
SMILES C(C(=O)O)N=C(N)N

Specifications

Color White
Quantity 25 g
Formula Weight 117.11
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Glycocyamine
EINECSNumber (9)-0329
TSCA No