accessibility menu, dialog, popup

UserName

OOHNH2NH2N

CAS RN 352-97-6

IUPAC Name: 2-[(diaminomethylidene)amino]acetic acid
Synonyms: β-guanidinoacetic acid
Molecular Weight (g/mol): 117.11
Molecular Formula: C3H7N3O2
InChi Key: BPMFZUMJYQTVII-UHFFFAOYSA-N
SMILES: NC(N)=NCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results