missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Glycocholic Acid, Sodium Salt, 100.1%, Calbiochem™,
$158.00 - $643.00
Chemical Identifiers
| CAS | 863-57-0 |
|---|---|
| Molecular Formula | C26H42NNaO6 |
| Molecular Weight (g/mol) | 487.61 |
| MDL Number | MFCD00036741 |
| InChI Key | OABYVIYXWMZFFJ-XIMBWSGONA-M |
| PubChem CID | 134129524 |
| IUPAC Name | sodium 2-[(4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]acetate |
| SMILES | [Na+].[H][C@@]1(CC[C@@]2([H])[C@]3([H])[C@H](O)C[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C)[C@H](C)CCC(=O)NCC([O-])=O |
Chemical Identifiers
| 863-57-0 | |
| 487.61 | |
| OABYVIYXWMZFFJ-XIMBWSGONA-M | |
| sodium 2-[(4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]acetate |
| C26H42NNaO6 | |
| MFCD00036741 | |
| 134129524 | |
| [Na+].[H][C@@]1(CC[C@@]2([H])[C@]3([H])[C@H](O)C[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C)[C@H](C)CCC(=O)NCC([O-])=O |
Specifications
| White | |
| 1 g | |
| C26H42NNaO6 | |
| OABYVIYXWMZFFJ-XIMBWSGONA-M | |
| sodium 2-[(4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]acetate | |
| 134129524 |
| 863-57-0 | |
| Glass Bottle | |
| MFCD00036741 | |
| [Na+].[H][C@@]1(CC[C@@]2([H])[C@]3([H])[C@H](O)C[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C)[C@H](C)CCC(=O)NCC([O-])=O | |
| 487.61 | |
| 100.1% (Titration)(dry basis) |