accessibility menu, dialog, popup

UserName

(R)(S)(S)(S)(R)(S)(S)(R)(R)NHOOO(R)OHOHCH3(R)CH3HOH3CHHHHHNa

CAS RN 863-57-0

IUPAC Name: sodium 2-[(4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]acetate
Synonyms: sodium glycocholate
Molecular Weight (g/mol): 487.61
Molecular Formula: C26H42NNaO6
InChi Key: OABYVIYXWMZFFJ-XIMBWSGONA-M
SMILES: [Na+].[H][C@@]1(CC[C@@]2([H])[C@]3([H])[C@H](O)C[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C)[C@H](C)CCC(=O)NCC([O-])=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results