missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Glycocholic Acid, Sodium Salt, 100.1%, Calbiochem™,

Catalog No. 3605121GM
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3605121GM 1 g Glass Bottle
3605125GM 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 3605121GM Supplier MilliporeSigma™ Supplier No. 3605121GM

Chemical Identifiers

CAS 863-57-0
Molecular Formula C26H42NNaO6
Molecular Weight (g/mol) 487.61
MDL Number MFCD00036741
InChI Key OABYVIYXWMZFFJ-XIMBWSGONA-M
PubChem CID 134129524
IUPAC Name sodium 2-[(4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]acetate
SMILES [Na+].[H][C@@]1(CC[C@@]2([H])[C@]3([H])[C@H](O)C[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C)[C@H](C)CCC(=O)NCC([O-])=O

Specifications

Color White
Quantity 1 g
Packaging Glass Bottle
Percent Purity 100.1% (Titration)(dry basis)