missing translation for 'onlineSavingsMsg'
Learn More

Glycine Sulfate 98.0+%, TCI America™

$292.65 - $728.84

Chemical Identifiers

CAS 513-29-1
Molecular Formula C6H17N3O10S
Molecular Weight (g/mol) 323.273
MDL Number MFCD00036426
InChI Key GZXOHHPYODFEGO-UHFFFAOYSA-N
Synonym glycine sulfate, triglycine sulfate, unii-neq4q0r0ya, neq4q0r0ya, triaminoacetic acid sulphate, 2-aminoacetic acid,sulfuric acid, 2-aminoacetic acid; sulfuric acid, tris glycine ; sulfuric acid
PubChem CID 10551
IUPAC Name 2-aminoacetic acid;sulfuric acid
SMILES C(C(=O)O)N.C(C(=O)O)N.C(C(=O)O)N.OS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
G04245G
View Documents
TCI America
G04245G
5 g N/A
Only null left
 
G042425G
View Documents
TCI America
G042425G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

513-29-1
323.273
GZXOHHPYODFEGO-UHFFFAOYSA-N
10551
C(C(=O)O)N.C(C(=O)O)N.C(C(=O)O)N.OS(=O)(=O)O
C6H17N3O10S
MFCD00036426
glycine sulfate, triglycine sulfate, unii-neq4q0r0ya, neq4q0r0ya, triaminoacetic acid sulphate, 2-aminoacetic acid,sulfuric acid, 2-aminoacetic acid; sulfuric acid, tris glycine ; sulfuric acid
2-aminoacetic acid;sulfuric acid

Specifications

513-29-1
C6H17N3O10S
5 g
GZXOHHPYODFEGO-UHFFFAOYSA-N
2-aminoacetic acid;sulfuric acid
10551
≥98.0% (N,T)
Glycine Sulfate
White
MFCD00036426
glycine sulfate, triglycine sulfate, unii-neq4q0r0ya, neq4q0r0ya, triaminoacetic acid sulphate, 2-aminoacetic acid,sulfuric acid, 2-aminoacetic acid; sulfuric acid, tris glycine ; sulfuric acid
C(C(=O)O)N.C(C(=O)O)N.C(C(=O)O)N.OS(=O)(=O)O
323.273
107.76
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'