CAS RN 513-29-1
IUPAC Name:
tris(2-aminoacetic acid); sulfuric acid
Synonyms:
triglycine sulfate
Molecular Weight (g/mol):
323.27
Molecular Formula:
C6H17N3O10S
InChi Key:
GZXOHHPYODFEGO-UHFFFAOYSA-N
SMILES:
NCC(O)=O.NCC(O)=O.NCC(O)=O.OS(O)(=O)=O
1
–
1
of
1
results
1
Glycine Sulfate 98.0+%, TCI America™
CAS: 513-29-1 Molecular Formula: C6H17N3O10S Molecular Weight (g/mol): 323.273 MDL Number: MFCD00036426 InChI Key: GZXOHHPYODFEGO-UHFFFAOYSA-N Synonym: glycine sulfate,triglycine sulfate,unii-neq4q0r0ya,neq4q0r0ya,triaminoacetic acid sulphate,2-aminoacetic acid,sulfuric acid,2-aminoacetic acid; sulfuric acid,tris glycine ; sulfuric acid PubChem CID: 10551 IUPAC Name: 2-aminoacetic acid;sulfuric acid SMILES: C(C(=O)O)N.C(C(=O)O)N.C(C(=O)O)N.OS(=O)(=O)O