missing translation for 'onlineSavingsMsg'
Learn More

Glycine Sulfate 98.0+%, TCI America™

Catalog No. G04245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G04245G 5 g
G042425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G04245G Supplier TCI America Supplier No. G04245G
Only null left

Chemical Identifiers

CAS 513-29-1
Molecular Formula C6H17N3O10S
Molecular Weight (g/mol) 323.273
MDL Number MFCD00036426
InChI Key GZXOHHPYODFEGO-UHFFFAOYSA-N
Synonym glycine sulfate, triglycine sulfate, unii-neq4q0r0ya, neq4q0r0ya, triaminoacetic acid sulphate, 2-aminoacetic acid,sulfuric acid, 2-aminoacetic acid; sulfuric acid, tris glycine ; sulfuric acid
PubChem CID 10551
IUPAC Name 2-aminoacetic acid;sulfuric acid
SMILES C(C(=O)O)N.C(C(=O)O)N.C(C(=O)O)N.OS(=O)(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 107.76
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Glycine Sulfate