missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine, Multi-Compendial, U.S.P., J.T. Baker™
$6,044.89 - $8,424.33
Chemical Identifiers
| CAS | 56-40-6 |
|---|---|
| Molecular Formula | C2H5NO2 |
| Molecular Weight (g/mol) | 75.07 |
| MDL Number | MFCD00008131 |
| InChI Key | DHMQDGOQFOQNFH-UHFFFAOYSA-N |
| Synonym | glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine |
| PubChem CID | 750 |
| ChEBI | CHEBI:15428 |
| IUPAC Name | 2-aminoacetic acid |
| SMILES | NCC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
02002950
|
Avantor J.T.Baker
058201 |
500 g | Wide Mouth Amber Glass Bottle |
Case of 4 Each for $8,424.33
|
|
||||
|
02002949
|
Avantor J.T.Baker
058205 |
2 kg | Wide Mouth Amber Glass Bottle |
Each for $6,044.89
|
|
||||
Chemical Identifiers
| C2H5NO2 | |
| MFCD00008131 | |
| glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
| CHEBI:15428 | |
| NCC(O)=O |
Specifications
| 56-40-6 | |
| MFCD00008131 | |
| DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
| 2-aminoacetic acid | |
| 750 | |
| 75.07 | |
| 1L = 1.16kg |
| C2H5NO2 | |
| glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
| NCC(O)=O | |
| 75.07 | |
| CHEBI:15428 | |
| Multi-Compendial/USP | |
| Glycine |