missing translation for 'onlineSavingsMsg'
Learn More

Glycine, Molecular biology reagent, ≥99%, MP Biomedicals™

Catalog No. ICN19482594
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19482594 5 kg
ICN19482580 100 g
ICN19482590 500 g
ICN19482591 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19482594 Supplier MP Biomedicals Inc Supplier No. 0219482594
Only null left
Protein Electrophoresis & Blotting

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Assay Percent Range ≥99%
Density 1.16g/mL (Lit.)
Quantity 5 kg
Formula Weight 75.07
Grade Molecular Biology
Physical Form Powder
Chemical Name or Material Glycine
EINECSNumber 200-272-2
Recommended Storage Store at room temperature (15° to 30°C).