missing translation for 'onlineSavingsMsg'
Learn More

Glycine Methyl Ester Hydrochloride 97.0+%, TCI America™

Catalog No. G024625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G024625G 25 g
G0246500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G024625G Supplier TCI America Supplier No. G024625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5680-79-5
Molecular Formula C3H8ClNO2
Molecular Weight (g/mol) 125.55
MDL Number MFCD00012870
InChI Key COQRGFWWJBEXRC-UHFFFAOYSA-N
Synonym glycine methyl ester hydrochloride, h-gly-ome.hcl, methyl 2-aminoacetate hydrochloride, methyl glycinate hydrochloride, glycine, methyl ester, hydrochloride, methyl glycinate hcl, methyl aminoacetate hydrochloride, glycine methyl ester hcl, glycine methyl ester hydrochloride salt, aminoacetic acid methyl ester hydrochloride
PubChem CID 122755
IUPAC Name hydrogen methyl 2-aminoacetate chloride
SMILES [H+].[Cl-].COC(=O)CN

Specifications

Melting Point 171°C
Color White
Quantity 25 g
Formula Weight 125.55
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Glycine Methyl Ester Hydrochloride
EINECSNumber (2)-1199
TSCA No