missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine, FCC, 98.5-101.5%, Spectrum™ Chemical
G1041, 56-40-6, C2H5NO2
$195.54 - $9,048.90
Chemical Identifiers
| CAS | 56-40-6 |
|---|---|
| Molecular Formula | C2H5NO2 |
| Molecular Weight (g/mol) | 75.07 |
| MDL Number | MFCD00008131 |
| InChI Key | DHMQDGOQFOQNFH-UHFFFAOYSA-N |
| IUPAC Name | 2-aminoacetic acid |
| SMILES | NCC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18603004
|
N/A | 100 g | Poly Bottle |
Each for $195.54
|
|
||||
|
18603005
|
N/A | 500 g | Poly Bottle |
Each for $424.56
|
|
||||
|
18603006
|
N/A | 2.5 kg | Poly Pail |
Each for $1,207.60
|
|
||||
|
18603007
|
N/A | 12 kg | Poly Pail |
Each for $3,556.10
|
|
||||
|
18603008
|
N/A | 50 kg | Fiber Drum |
Each for $9,048.90
|
|
||||