missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine, Electrophoresis Grade, MP Biomedicals™
Description
- Glycine is a component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis.
- Glycine is also a component of Towbin′s transfer buffer for Western blots.
- It is an inhibitory neurotransmitter in the spinal cord and an allosteric regulator of NMDA receptors.
Specifications
Specifications
| Melting Point | 451°F |
| pH | 4 (0.2 molar aq. soln.) |
| Specific Rotation | ≤0.0065% |
| Solubility Information | Soluble in water. Slightly soluble in alcohol and ethanol (95%). Almost insoluble in ether. |
| Density | 1.1607 g/mL |
| Quantity | 1 kg |
| Optical Rotation | ≤0.0065% |
| Percent Purity | ≥99.5% |
| Grade | Electrophoresis |
| Physical Form | Powder |
| Show More |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.