missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycine, Electrophoresis Grade, 99+%

Catalog No. p-7042920
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6436530 250 g
AAJ64365A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6436530 Supplier Thermo Scientific Chemicals Supplier No. J6436530
Only null left

CAS: 56-40-6 | C2H5NO2 | 75.07 g/mol

Glycine is used as a component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis. Glycine is also a component of Towbin?s transfer buffer for Western blots. Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine is used as a component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis. Glycine is also a component of Towbin′s transfer buffer for Western blots. Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Solubility
Freely soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Melting Point ∽245°C (decomposition)
pH 5.97
Color White
Linear Formula H2NCH2CO2H
Beilstein 635782
Merck Index 14,4491
Solubility Information Freely soluble in water
Density 1.595
Quantity 250 g
Odor Odorless
Formula Weight 75.07
Percent Purity ≥99.5%
Grade Electrophoresis
Physical Form Powder
Chemical Name or Material Glycine
Show More Show Less
EINECSNumber 200-272-2
RTECSNumber MB7600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only