Learn More
Glycine dimethylamide, 97%
CAS: 1857-19-8 | C4H10N2O | 102.137 g/mol
Supplier: Thermo Scientific Chemicals H5029303
Description
Glycine dimethylamide is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlycine dimethylamide is used as pharmaceutical intermediate.
Notes
Air sensitive. Incompatible with oxidizing agents and acids.
Specifications
| Glycine dimethylamide | |
| C4H10N2O | |
| MFCD06655265 | |
| 2-amino-n,n-dimethyl-acetamide, acetamide, 2-amino-n,n-dimethyl, n,n-dimethylglycinamide, glycinedimethyl amide, pubchem22320, acmc-1bw2m, 1,1-dimethylurea hydrochloride, 2-amino-n,n,-dimethylacetamide, 2-amino-n,n-dimethylacetamide; n,n-dimethylglycinamide | |
| CN(C)C(=O)CN | |
| 1 g | |
| 4961823 | |
| 102.14 | |
| 97% |
| 1857-19-8 | |
| H2NCH2CON(CH3)2 | |
| UN2735 | |
| KNVRBEGQERGQRP-UHFFFAOYSA-N | |
| 2-amino-N,N-dimethylacetamide | |
| 102.137 | |
| Amine-like | |
| Air Sensitive |
Chemical Identifiers
| 1857-19-8 | |
| MFCD06655265 | |
| 2-amino-n,n-dimethyl-acetamide, acetamide, 2-amino-n,n-dimethyl, n,n-dimethylglycinamide, glycinedimethyl amide, pubchem22320, acmc-1bw2m, 1,1-dimethylurea hydrochloride, 2-amino-n,n,-dimethylacetamide, 2-amino-n,n-dimethylacetamide; n,n-dimethylglycinamide | |
| 4961823 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only