Learn More
Glycine dimethylamide, 97%
CAS: 1857-19-8 | C4H10N2O | 102.137 g/mol
$305.40 - $305.40
Chemical Identifiers
| CAS | 1857-19-8 |
|---|---|
| MDL Number | MFCD06655265 |
| Synonym | 2-amino-n,n-dimethyl-acetamide, acetamide, 2-amino-n,n-dimethyl, n,n-dimethylglycinamide, glycinedimethyl amide, pubchem22320, acmc-1bw2m, 1,1-dimethylurea hydrochloride, 2-amino-n,n,-dimethylacetamide, 2-amino-n,n-dimethylacetamide; n,n-dimethylglycinamide |
| PubChem CID | 4961823 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5029303
|
Thermo Scientific Chemicals
H5029303 |
1 g |
Each for $305.40
|
|
|||||
Description
Glycine dimethylamide is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlycine dimethylamide is used as pharmaceutical intermediate.
Notes
Air sensitive. Incompatible with oxidizing agents and acids.
Chemical Identifiers
| 1857-19-8 | |
| MFCD06655265 | |
| 2-amino-n,n-dimethyl-acetamide, acetamide, 2-amino-n,n-dimethyl, n,n-dimethylglycinamide, glycinedimethyl amide, pubchem22320, acmc-1bw2m, 1,1-dimethylurea hydrochloride, 2-amino-n,n,-dimethylacetamide, 2-amino-n,n-dimethylacetamide; n,n-dimethylglycinamide | |
| 4961823 |
Specifications
| 1857-19-8 | |
| H2NCH2CON(CH3)2 | |
| UN2735 | |
| KNVRBEGQERGQRP-UHFFFAOYSA-N | |
| 2-amino-N,N-dimethylacetamide | |
| 102.137 | |
| Amine-like | |
| Air Sensitive | |
| Glycine dimethylamide |
| C4H10N2O | |
| MFCD06655265 | |
| 2-amino-n,n-dimethyl-acetamide, acetamide, 2-amino-n,n-dimethyl, n,n-dimethylglycinamide, glycinedimethyl amide, pubchem22320, acmc-1bw2m, 1,1-dimethylurea hydrochloride, 2-amino-n,n,-dimethylacetamide, 2-amino-n,n-dimethylacetamide; n,n-dimethylglycinamide | |
| CN(C)C(=O)CN | |
| 1 g | |
| 4961823 | |
| 102.14 | |
| 97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only