missing translation for 'onlineSavingsMsg'
Learn More

Glycine Crystalline MP Biomedicals

Catalog No. ICN10057025
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10057025 25 kg
ICN10057001 1 kg
ICN10057005 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10057025 Supplier MP Biomedicals Inc Supplier No. 0210057025
Only null left

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Density 1.16 g/mL
Quantity 25 kg
Formula Weight 75.07
Physical Form Crystalline
Chemical Name or Material Glycine