missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine, Crystalline, MP Biomedicals
Specifications
Specifications
| Quantity | 100 g |
| Formula Weight | 75.07 |
| Grade | Cell Culture |
| Physical Form | Crystals |
| Chemical Name or Material | Glycine, Free Acid |
Safety and Handling
| Recommended Storage | Room temperature |