Learn More
Glycine (Crystalline Granules or Powder/USP), Fisher Chemical™
Supplier: Fisher Chemical FLG4812
Specifications
| 182°C to 236°C | |
| 56-40-6 | |
| White | |
| Pass Test (USP) | |
| Pass Test (USP) | |
| C2H5NO2 | |
| CH2(NH2)(COOH) | |
| Meets Requirements (USP) | |
| glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
| 1.595g/cm³ | |
| 2-aminoacetic acid | |
| 12 kg | |
| CHEBI:15428 | |
| 98.5 to 101.5% (USP) | |
| Powder |
| Glycine | |
| 5.9 to 6.4 | |
| 0.007% max. (USP) | |
| 0.1% max. (USP) | |
| 0.2% max. (USP) | |
| Poly Pail | |
| MFCD00008131 | |
| 0.0065% max. (USP) | |
| DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
| NCC(O)=O | |
| 75.07 | |
| 750 | |
| 75.07 | |
| USP |
Chemical Identifiers
| 56-40-6 | |
| 75.07 | |
| DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
| 750 | |
| 2-aminoacetic acid |
| C2H5NO2 | |
| MFCD00008131 | |
| glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
| CHEBI:15428 | |
| NCC(O)=O |
Safety and Handling
CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .
NFPA
Health:1
Flammability:1
Instability:1