Learn More
Glycine (Crystalline Granules or Powder/USP/EP/BP/JP), Fisher Chemical™
Supplier: Thermo Fisher Scientific G4512
Specifications
| 182°C to 236°C | |
| 0.02% max. (JP) | |
| 2ppm max. (JP) | |
| White | |
| 0.007% max. (USP), 0.021% max. (JP), 75ppm max. (BP), 75ppm max. (BP/EP), 0.021% max. (JP), 0.007% max. (USP), 75ppm max. (EP) | |
| Pass Test (USP) | |
| Pass Test (BP), Pass Test (BP/EP/JP/USP), Pass Test (EP), Pass Test (JP), Pass Test (USP) | |
| Pass Test (JP) | |
| Poly Pail | |
| MFCD00008131 | |
| 0.10% max. (JP) | |
| 0.1% max. (BP), 0.1% max. (BP/EP), 0.1% max. (EP) | |
| DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
| NCC(O)=O | |
| 75.07 | |
| 750 | |
| 75.07 | |
| BP/EP/JP/USP |
| Glycine | |
| 56-40-6 | |
| 5.9 to 6.4 (BP/EP/JP) | |
| Report EU/g (EP) | |
| 10ppm max. (BP), 10ppm max. (BP/EP), 20ppm max. (JP), 10ppm max. (EP), 20ppm max. (JP) | |
| 0.1% max. (USP) | |
| 0.2% max. (USP), 0.30% max. (JP), 0.5% max. (BP), 0.5% max. (BP/EP), 0.30% max. (JP), 0.2% max. (USP), 0.5% max. (EP) | |
| C2H5NO2 | |
| CH2(NH2)(COOH) | |
| Meets Requirements (BP), Meets Requirements (BP/EP/USP), Meets Requirements (EP), Meets Requirements (USP) | |
| 0.0065% max. (USP), 0.028% max. (JP), 0.028% max. (JP), 0.0065% max. (USP) | |
| glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
| 1.595g/cm³ | |
| 2-aminoacetic acid | |
| 12 kg | |
| CHEBI:15428 | |
| 98.5 to 101.0% (EP) | |
| Powder |
Chemical Identifiers
| 56-40-6 | |
| 75.07 | |
| DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
| 750 | |
| 2-aminoacetic acid |
| C2H5NO2 | |
| MFCD00008131 | |
| glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
| CHEBI:15428 | |
| NCC(O)=O |
Safety and Handling
CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .
NFPA
Health:1
Flammability:1
Instability:1