missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycine, ACS, 98.5+%

Catalog No. AA43497A5
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43497A5 3 kg
AA4349722 100 g
AA4349736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA43497A5 Supplier Thermo Scientific Chemicals Supplier No. 043497A5
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56-40-6 | C2H5NO2 | 75.07 g/mol

Glycine is employed to protect the kidneys from the harmful side effects of certain drugs used after organ transplantation as well as the liver from the harmful effects of alcohol. The solution of glycine plays an important role for deposition of alloys and metals. It is likewise useful for patients with acute alcohol hallucinosis. It is applied for treating schizophrenia, stroke, benign prostatic hyperplasia (BPH), and some rare inherited metabolic disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Melting Point ∼245°C (decomposition)
pH 5.97
Color White
Beilstein 635782
Merck Index 14,4491
Solubility Information Soluble in pyridine,water. Sparingly soluble in ethanol. Insoluble in ether.
Density 1.595 g/mL
Quantity 3 kg
Odor Odorless
Formula Weight 75.07
Percent Purity ≥98.5%
Grade ACS
Physical Form Crystalline
Chemical Name or Material Glycine
Show More Show Less
EINECSNumber 200-272-2
RTECSNumber MB7600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only