missing translation for 'onlineSavingsMsg'
Learn More

Glycine, ≥99%, Molecular Biology Grade, Ultrapure

Catalog No. AAJ16407A7
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ16407A7 5 kg Plastic Bottle
AAJ1640736 500 g Plastic Bottle
AAJ16407A1 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ16407A7 Supplier Thermo Fisher Scientific Supplier No. J16407A7

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56-40-6 | C2H5NO2 | 75.067 g/mol

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.067
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES C(C(=O)O)N

Specifications

Melting Point ∼245°C (decomposition)
Packaging Plastic Bottle
Linear Formula H2NCH2CO2H
Beilstein 635782
Density 1.595
Quantity 5 kg
Formula Weight 75.07
Percent Purity ≥99%
Grade Molecular Biology
Specific Gravity 1.595
Physical Form Powder
Chemical Name or Material Glycine
Show More Show Less
EINECSNumber 200-272-2
RTECSNumber MB7600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only