missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine, ≥99%, Molecular Biology Grade, Ultrapure
Specifications
Specifications
| Melting Point | ∼245°C (decomposition) |
| Packaging | Plastic Bottle |
| Linear Formula | H2NCH2CO2H |
| Beilstein | 635782 |
| Density | 1.595 |
| Quantity | 1 kg |
| Formula Weight | 75.07 |
| Percent Purity | ≥99% |
| Grade | Molecular Biology |
| Specific Gravity | 1.595 |
| Show More |
Safety and Handling
| EINECSNumber | 200-272-2 |
| RTECSNumber | MB7600000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only